Skip to Content
Merck
Cobalt(II) carbonate hydrate ≥99.99% trace metals basis

Cobalt(II) carbonate hydrate

Cobalt(II) carbonate hydrate ≥99.99% trace metals basis
Linear Formula:
CoCO3·xH2O
CAS Number:
Molecular Weight:
118.94 (anhydrous basis)

Products

Product Category
Formula Weight
Physical Form
Purity
Chemical Composition
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing