Citalopram Related Compound A
Synonym(s):
1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarboxamide
Empirical Formula (Hill Notation):
C20H23FN2O2
CAS Number:
Molecular Weight:
342.41
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2