
Ciclopirox olamine

Synonym(s):
6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridone ethanolammonium salt
Empirical Formula (Hill Notation):
C12H17NO2 · C2H7NO
CAS Number:
Molecular Weight:
268.35
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing