
Chlorthalidone Related Compound A

Synonym(s):
4′-Chloro-3′-sulfamoyl-2-benzophenone-2-carboxylic acid, 2-[3-(Aminosulfonyl)-4-chlorobenzoyl]benzoic acid
Empirical Formula (Hill Notation):
C14H10ClNO5S
CAS Number:
Molecular Weight:
339.75
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2