Skip to Content
BM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon, mesoporous
Empirical Formula (Hill Notation):
C
CAS Number:
1333-86-4
Molecular Weight:
12.01
Products
Product Category
anode materials (4)
Brand
Sigma-Aldrich (5)
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Application
Particle Size (µm)
Pore Size (Å)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
699632
Carbon, mesoporous,
nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Expand
View Pricing
699624
Carbon, mesoporous,
nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Expand
View Pricing
699640
Carbon, mesoporous,
less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Expand
View Pricing
702102
Carbon, mesoporous,
Expand
View Pricing
702110
Carbon, mesoporous,
hydrophilic pore surface
Expand
View Pricing