![Carbendazim certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/280/526/09ce670c-19ab-45ce-83af-19f8e5b408db/640/09ce670c-19ab-45ce-83af-19f8e5b408db.png)
Carbendazim
![Carbendazim certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/280/526/09ce670c-19ab-45ce-83af-19f8e5b408db/640/09ce670c-19ab-45ce-83af-19f8e5b408db.png)
Synonym(s):
BCM, Methyl 2-benzimidazolecarbamate, Methyl benzimidazol-2-ylcarbamate
Empirical Formula (Hill Notation):
C9H9N3O2
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3