
Boc-D-Glu-OBzl

Synonym(s):
Boc-D-glutamic acid 1-benzyl ester, Boc-D-Glu-OBzl
Empirical Formula (Hill Notation):
C17H23NO6
CAS Number:
Molecular Weight:
337.37
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing