
Bixafen

Synonym(s):
N-(3′,4′-Dichloro-5-fluorobiphenyl-2-yl)-3-(difluoromethyl)-1-methylpyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C18H12Cl2F3N3O
CAS Number:
Molecular Weight:
414.21
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2