
β-Estradiol-6-one

Synonym(s):
1,3,5-Estratriene-3,17β-diol-6-one, 3,17β-Dihydroxy-1,3,5(10)-estratrien-6-one, 6-Ketoestradiol
Empirical Formula (Hill Notation):
C18H22O3
CAS Number:
Molecular Weight:
286.37
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing