
Benzaldehyde propylene glycol acetal

Synonym(s):
Benzaldehyde propylene glycol acetal, mixture of isomers, 4-Methyl-2-phenyl-1,3-dioxolane
Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2