
Baicalein

Synonym(s):
5,6,7-Trihydroxyflavone, Baicalein
Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade