
Apomorphine hydrochloride

Synonym(s):
R-(−)-Apomorphine hydrochloride hemihydrate
Linear Formula:
C17H17NO2 · HCl · 1/2H2O
CAS Number:
Molecular Weight:
312.79
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2