
Anti-PLGRKT antibody produced in rabbit

Synonym(s):
Anti-C9orf46, Anti-Uncharacterized protein C9orf46 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1