
Anti-Avidin–FITC antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-Avidin
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing