Skip to Content
Merck
Ammonium Hydroxide OmniTrace® Ultra

Ammonium Hydroxide

Ammonium Hydroxide OmniTrace® Ultra
Synonym(s):
Ammonium hydroxide solution, Ammonia aqueous, Ammonia water
Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05

Products

Product Category
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Special Grade