Skip to Content
BM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium aluminum sulfate dodecahydrate
Synonym(s):
Aluminum ammonium sulfate dodecahydrate, Ammonium alum
Linear Formula:
AlNH
4
(SO
4
)
2
· 12H
2
O
CAS Number:
7784-26-1
Molecular Weight:
453.33
EC Number:
232-055-3
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
A2140
Ammonium aluminum sulfate dodecahydrate,
ReagentPlus
®
, ≥99% (titration)
Expand
View Pricing