
Amlodipine Related Compound C

Synonym(s):
Dimethyl 4-(2-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Empirical Formula (Hill Notation):
C17H18ClNO4
CAS Number:
Molecular Weight:
335.78
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2