
Adenosine 5′-diphosphate monopotassium salt dihydrate

Synonym(s):
ADP
Empirical Formula (Hill Notation):
C10H14KN5O10P2 · 2H2O
CAS Number:
Molecular Weight:
501.32
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2