Skip to Content
Merck
9-Bromo-1-nonanol 95%

9-Bromo-1-nonanol

9-Bromo-1-nonanol 95%
Synonym(s):
Nonamethylene bromohydrin
Linear Formula:
Br(CH2)9OH
CAS Number:
Molecular Weight:
223.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing