
7-Hydroxyflavone

Synonym(s):
7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, NSC 94258
Empirical Formula (Hill Notation):
C15H10O3
CAS Number:
Molecular Weight:
238.24
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing