Skip to Content
Merck
7-Fluoroisatin 96%

7-Fluoroisatin

7-Fluoroisatin 96%
Synonym(s):
7-Fluoroindole-1H-2,3-dione
Empirical Formula (Hill Notation):
C8H4FNO2
CAS Number:
Molecular Weight:
165.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing