Skip to Content
Merck
5-Methyl-3-heptanone ≥94% (GC)

5-Methyl-3-heptanone

5-Methyl-3-heptanone ≥94% (GC)
Synonym(s):
Ethyl isoamyl ketone, 5-Methyl-3-heptanone
Linear Formula:
CH3CH2COCH2CH(CH3)CH2CH3
CAS Number:
Molecular Weight:
128.21

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing