4-Hydroxybenzylamine
Synonym(s):
4-(Aminomethyl)phenol
Empirical Formula (Hill Notation):
C7H9NO
CAS Number:
Molecular Weight:
123.15
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing