
2-Mercaptoethanol

Synonym(s):
β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol
Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Products
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing