
2-Chlorotrityl chloride

Synonym(s):
2-Chlorotrityl chloride, Chloro(2-chlorophenyl)diphenylmethane
Empirical Formula (Hill Notation):
C19H14Cl2
CAS Number:
Molecular Weight:
313.22
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3