
2-Acetamido-2-deoxy-β-D-glucopyranosyl azide

Synonym(s):
2-Acetamido-1-azido-1,2-dideoxy-β-D-glucopyranose
Empirical Formula (Hill Notation):
C8H14N4O5
CAS Number:
Molecular Weight:
246.22
Products
Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Optical Isomer
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing