Skip to Content
Merck
1-(Chloromethyl)-1-methyl-4-(2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-yl)piperazin-1-ium chloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1-(Chloromethyl)-1-methyl-4-(2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-yl)piperazin-1-ium chloride

1-(Chloromethyl)-1-methyl-4-(2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-yl)piperazin-1-ium chloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
N-Chloromethyl-olanzapine chloride
Empirical Formula (Hill Notation):
C18H22Cl2N4S
CAS Number:
Molecular Weight:
397.37

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade