Skip to Content
Merck
1,4-Diethylbenzene ≥98%

1,4-Diethylbenzene

1,4-Diethylbenzene ≥98%
Synonym(s):
1,4-Diethylbenzene
Linear Formula:
C6H4(C2H5)2
CAS Number:
Molecular Weight:
134.22

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing