Skip to Content
Merck
1,3-Bis(2,6-di(3-pentyl)phenyl)imidazolium chloride ≥95%

1,3-Bis(2,6-di(3-pentyl)phenyl)imidazolium chloride

1,3-Bis(2,6-di(3-pentyl)phenyl)imidazolium chloride ≥95%
Empirical Formula (Hill Notation):
C35H53ClN2
CAS Number:
Molecular Weight:
537.26

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing