
12-O-Methylcarnosic acid

Synonym(s):
12-Methoxycarnosic acid, Carnosic acid 12-methyl ether
Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing