
1,2-Epoxybutane

Synonym(s):
α-Butylene oxide, 1,2-Butylene oxide, 1-Butene oxide, Ethyloxirane, 1,2-Epoxybutane
Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing