![1,2,4-Trimethylbenzene 98%](/deepweb/assets/sigmaaldrich/product/structures/917/596/1c2896ac-ed29-446c-82be-470bbf87d530/640/1c2896ac-ed29-446c-82be-470bbf87d530.png)
1,2,4-Trimethylbenzene
![1,2,4-Trimethylbenzene 98%](/deepweb/assets/sigmaaldrich/product/structures/917/596/1c2896ac-ed29-446c-82be-470bbf87d530/640/1c2896ac-ed29-446c-82be-470bbf87d530.png)
Synonym(s):
Pseudocumene, 1,2,4-Trimethylbenzene
Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing