
11-Ketotestosterone

Synonym(s):
17β-Hydroxy-4-androstene-3,11-dione, 17β-Hydroxyandrost-4-ene-3,11-dione, 4-Androsten-17β-ol-3,11-dione
Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing