Skip to Content
Merck
All Photos(1)

Key Documents

G006123

HPLC Analysis of Morphine and Glucuronide Metabolites on Discovery® HS F5

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Discovery HS F5, 5 cm x 4.6 mm I.D., 5 μm particles (567513-U)
mobile phase: [A] 10 mM ammonium acetate, pH 6.8 (unadjusted); [B] acetonitrile
gradient: 0% B for 1 min, to 30% B in 4 min, held 3 min
flow rate: 1 mL/min
column temp.: 40 °C
detector: UV, 280 nm
injection: 10 μL
sample: 50 μg/mL in final mobile phase

suitability

application for HPLC

application(s)

clinical
pharmaceutical (small molecule)

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Morphine 3-ß-D-glucuronide
    Morphine-6-β-D-glucuronide solution, 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®
    Morphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service