Skip to Content
Merck
All Photos(1)

Key Documents

CRM69121

Supelco

Diphenylamine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NH
CAS Number:
Molecular Weight:
169.22
Beilstein:
508755
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

vapor density

5.82 (vs air)

vapor pressure

1 mmHg ( 108 °C)

product line

TraceCERT®

autoignition temp.

1175 °F

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

bp

302 °C (lit.)

mp

50-53 °C (lit.)

format

neat

SMILES string

N(c1ccccc1)c2ccccc2

InChI

1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H

InChI key

DMBHHRLKUKUOEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Diphenylamine is a fat-soluble organic antioxidant, widely used to control post-harvest skin scalding in apples and pears by preventing the oxidation of α-farnesene.
According to the Commission Implementing Regulation (EU) No 578/2012 of 29th June 2012, diphenylamine is not approved for use in the European Union (EU) as an active substance in the plant protection products, following the Regulation (EC) No. 1107/2009. But a default maximum residue limit of 0.05 mg/kg is allowed for its presence in the products of plant and animal origin as per the EU Reg. 2018/1515.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Diphenylamine CRM may also be used as given below:

  • Development and validation of a gas chromatography-mass spectrometry (GC-MS) method for the simultaneous analysis of diphenylamine, tolylfluanid, propargite, and phosalone in human liver samples
  • Determination of diphenylamine residues in fruit samples after applying a combination of supercritical fluid extraction (SFE) and supramolecular solvent-based microextraction (SSME) for the extraction, followed by a determination using high-performance liquid chromatography (HPLC) with ultraviolet (UV) detection
  • Evaluate a newly designed molecularly imprinted polymer (MIP) based on β-cyclodextrin (β -CD) and graphene oxide (GO) for the determination of diphenylamine
  • Analysis of diphenylamine in apple samples using an electrochemical sensor based on phosphomolybdic acid (PMo12) and graphene oxide (GO)
  • Investigate the performance of an electrochemical sensor founded upon polymeric carbon-rich graphitic carbon nitride (C-gCN) to detect diphenylamine in apple juice samples
  • Study the determination of diphenylamine in apple and pear samples using an electrochemical sensor based on Copper@nanoporous carbon (Cu@NPC) as the electrode material

Other Notes

Ion Mobility: TWCCSN2 value of 135.5 Å2 [M+H]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N CRM69121 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney,Liver,spleen

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service