Skip to Content
Merck
All Photos(1)

Key Documents

T8700

Sigma-Aldrich

Anti-Tyrosine Hydroxylase antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution, sufficient for 10 blots

Synonym(s):

Rabbit Anti-TYH

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

usage

sufficient for 10 blots

species reactivity

mammals

technique(s)

immunohistochemistry (frozen sections): 1:1,000
indirect immunofluorescence: 1:1,000
western blot: 1:1,000 using rat brain lysates of PC-12 cells stimulated by okadaic acid

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TH(7054)
mouse ... Th(21823)
rat ... Th(25085)

General description

Tyrosine hydroxylase (TH) is encoded by the gene mapped to human chromosome 11p15. TH is a tetramer of four identical subunits, which is characterized with a regulatory, catalytic, and tetramerization domains. The enzyme utilizes tyrosine, tetrahydrobiopterin (BH4) and O2 as cosubstrates, and Fe2+ as a cofactor.

Immunogen

SDS-denatured rat tyrosine hydroxylase purified from pheochromocytoma.

Application

Anti-tyrosine hydroxylase antibody produced in rabbit has been used in western blot analysis and immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Tyrosine hydroxylase (TH) catalyzes the first rate limiting step in the biosynthesis of catecholamine neurotransmitter i.e. the conversion of L-tyrosine to L-dopa. Inhibition of TH by L-phenylalanine, might play a crucial role in phenylketonuria and block the synthesis of norepinephrine. Activity of TH can be regulated by phosphorylation. Decreased expression of TH is associated with various neuropsychiatric diseases such as schizophrenia and Parkinson′s disease (PD).

Target description

Tyrosine hydroxylase (TH) is involved in the conversion of phenylalanine to dopamine. It catalyzes the initial, rate-limiting step of the catecholamine biosynthetic pathway.

Physical form

Solution in 100 μL in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge Mendoza et al.
Journal of neurochemistry, 112(6), 1489-1499 (2010-01-14)
Under special restricted feeding conditions the mammalian circadian clock, contained in the hypothalamic suprachiasmatic nucleus (SCN), can be entrained by food. During food restriction, hungry animals are very motivated to obtain food. This motivational state could be a key component
Maheep Bhatnagar et al.
PloS one, 12(5), e0177254-e0177254 (2017-05-17)
Parkinson's disease remains as one of the most common debilitating neurodegenerative disorders. With the hopes of finding agents that can cure or reduce the pace of progression of the disease, we studied two traditional medicinal plants: Centella asiatica and Withania
Z Jallah et al.
BJOG : an international journal of obstetrics and gynaecology, 123(7), 1076-1085 (2015-08-25)
To evaluate the impact of prolapse meshes on vaginal smooth muscle structure (VaSM) and function, and to evaluate these outcomes in the context of the mechanical and textile properties of the mesh. Three months following the implantation of three polypropylene
Chi-Lin Li et al.
Nutrients, 11(7) (2019-07-25)
Schisandra chinensis (Turcz.) Baill. (S. chinensis) is a well-known botanical medicine and nutritional supplement that has been shown to have potential effects on neurodegeneration. To investigate the potential neuroprotective effect of S. chinensis fruit extract, 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) was used to
Yanqin Wang et al.
PPAR research, 2017, 4089214-4089214 (2017-03-31)
The aim of the present study was to gain insight into the neuroprotection effects and mechanism of thiazolidinedione pioglitazone in both in vitro and in vivo MPP+/MPTP induced PD models. In vivo experimental results showed that oral treatment of pioglitazone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service