Skip to Content
Merck
All Photos(1)

Key Documents

SML1152

Sigma-Aldrich

GSK137647A

≥98% (HPLC)

Synonym(s):

4-Methoxy-N-(2,4,6-trimethylphenyl)-benzenesulfonamide, N-Mesityl-4-methoxybenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19NO3S
CAS Number:
Molecular Weight:
305.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(NS(C2=CC=C(OC)C=C2)(=O)=O)C(C)=CC(C)=C1

InChI

1S/C16H19NO3S/c1-11-9-12(2)16(13(3)10-11)17-21(18,19)15-7-5-14(20-4)6-8-15/h5-10,17H,1-4H3

InChI key

FQUAFMNPXPXOJE-UHFFFAOYSA-N

Biochem/physiol Actions

GSK137647A is a selective agonist of the free fatty acid receptor 4 (FFA4/GPR120), which is involved in long-chain fatty acid stimulated GLP-1 secretion and has been implicated in glucose homeostasis, adiposity, and regulation of the anti-inflammatory response. GSK137647A has pEC50 values of 6.3, 6.2, and 6.1 for human, mouse and rat FFA4, and pEC50 values < 4.5 for all three species for FFA1, FFA2, and FFA3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service