Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9030-59

Ab Buffer Kit

Cytiva 28-9030-59, pack of 1 kit

Synonym(s):

Ab Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.25

sterility

non-sterile

packaging

pack of 1 kit

manufacturer/tradename

Cytiva 28-9030-59

storage condition

(20% Ethanol)

storage temp.

2-8°C

General description

Ab Buffer Kit contains carefully prepared buffer concentrates for binding, washing, and elution of IgG according to recommended protocols. The kit also includes neutralizing buffer. Includes: 1 × 50 ml Binding buffer 10× stock solution, 1 × 15 ml Elution buffer 10× stock solution, and 1 × 25 ml Neutralizing buffer.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to separate IgG antibodies by affinity chromatography using HiTrap® Protein G HP columns from Cytiva.

This page shows various purification options for Protein A Sepharose chromatography media and describes typical binding and elution conditions for Protein A Sepharose chromatography media.

Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.

Protocols

This page shows how to separate IgG antibodies by affinity chromatography using Protein G HP SpinTrap™/Ab SpinTrap™ from Cytiva.

This page shows how to separate IgG antibodies by affinity chromatography using Protein G GraviTrap from Cytiva.

This page shows how to separate IgG antibodies by affinity chromatography using Protein G GraviTrap from Cytiva.

This page shows how to separate IgG antibodies by affinity chromatography using Protein G GraviTrap from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service