Key Documents
GE17-0572-02
Sephadex® G-25 DNA GR
Cytiva 17-0572-02, pack of 100 g
About This Item
UNSPSC Code:
23151817
NACRES:
NA.52
packaging
pack of 100 g
manufacturer/tradename
Cytiva 17-0572-02
storage condition
(.01 M NaOH)
particle size
20-80 μm (dry)
working range
2-13
Related Categories
General description
illustra Sephadex® G-25 DNA grade SF.
Features and Benefits
  • For purification of DNA ( 10 bases in length) from small molecules by gel filtration.
  • Tested to ensure reproducibly high recovery of DNA.
  • Used in MicroSpin G-25 Columns and NAP Columns.
  • Enables self preparation of columns for nucleic acid purification.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
MicroSpin is a trademark of Cytiva
NAP is a trademark of Cytiva
Sephadex is a registered trademark of Cytiva
illustra is a trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Visit the Document Library
Articles
This page shows how to solve practical problems that may occur when running an affinity chromatography column.
Protocols
This page shows describes fractional precipitation for removing gross impurities from affinity chromatography samples with small volumes.
This page shows how to perform a separation with a Sephadex column from Cytiva which are Size Exclusion Chromatography media consisting of an epichlorohydrin cross-linked dextran.
How to perform buffer exchange and desalting with Sephadex G-25, HiTrap Desalting columns, or ÄKTAprime plus.
This page shows how to prepare samples for purification with affinity chromatography.
Related Content
Separation options for Size Exclusion Chromatography with Sephadex from Cytiva.