Key Documents
EP5895155006
Rotor for Eppendorf® 5910 R Centrifuge
FA-30x2, incl. aerosol-tight QuickLock Rotor for Eppendorf(R) lid, for 30 x 2 mL tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 30 x 1.5/2.0 mL tubes
FA-30x2, incl. aerosol-tight QuickLock Rotor for Eppendorf(R) lid, for 30 x 2 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895155006
parameter
13,700 max. RCF (20,984 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5910R
General description
Rotor for Eppendorf(R)5910R Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG