Skip to Content
Merck
All Photos(1)

Key Documents

T84 Cell Line human

88021101, human colon (lung metastasis), Epithelial

Synonym(s):

T-84 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

T84 Cell Line human, 88021101

biological source

human colon (lung metastasis)

description

Human colon carcinoma

growth mode

Adherent

karyotype

2n = 46, modal no. 56

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon carcinoma

Cell Line Description

Derived from a lung metastasis of colon carcinoma in a 72 year old male. Tumour tissue was inoculated sub-cutaneously and serially transplanted in BALB/c nude mice and subsequently established in in vitro culture. The histological characteristics of the tumour were maintained throughout the transplantation procedure. They have receptors for a range of peptide hormones and neurotransmitters. The cells grow as monolayers, exhibit tight junctions and desmosomes between adjacent cells.

Application

Human T84 cell line has been used for comparative genome analysis of Salmonella enterica serovar Typhimurium definitive type 2 (DT2) isolate 94-213 and S. Typhimurium isolates SL1344, DT104 and D23580. The ability of these isolates to invade culture medium has also been studied. It has also been used to construct and study the suitability of an in vitro model to study chemotherapy-induced mucosal damage.
Tumourigenicity, neurotransmitter and hormone studies

Culture Medium

Ham′s F12 + DMEM (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 - 1:4 i.e. seeding at 1-3 x 10,000 cells / cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Cells grow slowly and may not form a complete monolayer, maintain at high density with a minimum of 25% confluency.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service