Skip to Content
Merck
All Photos(1)

Key Documents

MiCL1 (S+L-)

86020603, mink lung, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MiCL1 (S+L-), 86020603

biological source

mink lung

growth mode

Adherent

karyotype

2n = 30

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mink lung, Moloney murine sarcoma virus infected

Cell Line Description

A clone of the mink lung cell line Mv.1.Lu (Sigma Catalogue number. 88050503). Isolated from a single cell of a focus produced in the monolayer after infection with Moloney murine sarcoma virus associated with the RD-114 helper leukaemia virus. Super infection with non-transforming leukaemia virus results in transformation and net amplification of sarcoma-specific RNA sequences.

Application

Virus studies: Moloney MSV and RD-114 helper leukaemia virus Leukemogenesis

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service