Skip to Content
Merck
All Photos(1)

Documents

EL4.BU.OU6

85121301, mouse blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

EL4.BU.OU6, 85121301

biological source

mouse blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Interleukin 2 (IL-2), Interleukin 4 (IL-4), Thy-1.2

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C57BL/6N ascites lymphoma lymphoblast

Cell Line Description

Derivative of EL4.BU. Resistant to BUdR and ouabain and expresses Thy-1.2 Ag. Will die in HAT selective medium. A high producer of interleukin 2 and interleukin 4.

Application

Resistance and hormone studies

Culture Medium

DMEM + 2mM Glutamine + 0.05mM 2-Mercaptoethanol (2ME) + 1.16g/l 1% L-Arginine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service