Skip to Content
Merck
All Photos(1)

Key Documents

C5488

Sigma-Aldrich

Anti-Calcium Channel (α1G Subunit) (T-type Voltage-Gated Ca2+ Channel) antibody produced in rabbit

~1 mg/mL, fractionated antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

western blot: 5-10 μg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Specificity

The antibody specifically recognizes a splice variant of α1G.

Immunogen

synthetic peptide derived from the rat α1G calcium channel subunit conjugated to KLH.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuyako Ohkubo et al.
International journal of oncology, 41(1), 267-275 (2012-04-04)
T-type voltage-gated Ca2+ channels have unique electrophysiological properties, suitable for generating Ca2+ oscillations and waves and thus controlling the proliferation of various tumor cells. In the present study, we investigated the role of Cav3.1, a candidate tumor suppressor gene, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service