Skip to Content
Merck
All Photos(1)

Key Documents

S1250000

Stanozolol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

17β-Hydroxy-17α-methyl-5α-androstano[3,2-c]pyrazole, 5α-Androstane-17α-methyl-17β-ol-[3,2-c]pyrazole, Androstanazole, Stanazol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H32N2O
CAS Number:
Molecular Weight:
328.49
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

stanozolol

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CC[C@]3([H])[C@]([H])(CC[C@@]4(C)[C@@]3([H])CC[C@]4(C)O)[C@@]1(C)Cc5cn[nH]c5C2

InChI

1S/C21H32N2O/c1-19-11-13-12-22-23-18(13)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,24H,4-11H2,1-3H3,(H,22,23)/t14-,15+,16-,17-,19-,20-,21-/m0/s1

InChI key

LKAJKIOFIWVMDJ-IYRCEVNGSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Stanozolol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Helfman et al.
Journal of the American Academy of Dermatology, 33(2 Pt 1), 254-258 (1995-08-01)
Anabolic steroids are synthetic derivatives of testosterone that were developed in the 1950s in an attempt to dissociate the anabolic and androgenic effects of testosterone. The anabolic steroid stanozolol has been particularly helpful because it has one of the largest
Maya K Nanda et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 113(2), 198-203 (2014-06-15)
Current guidelines recommend short-term prophylaxis (STP) before invasive procedures to prevent hereditary angioedema (HAE) attacks; however, adherence to these guidelines may be variable because this indication lacks Food and Drug Administration approval in the United States. To ascertain the STP
David W Andersen et al.
Journal of analytical toxicology, 38(9), 637-644 (2014-08-12)
A screening method for 18 frequently measured exogenous anabolic steroids and the testosterone/epitestosterone (T/E) ratio in forensic cases has been developed and validated. The method involves a fully automated sample preparation including enzyme treatment, addition of internal standards and solid
Nathaly Reyes-Garcés et al.
Journal of chromatography. A, 1374, 40-49 (2014-12-03)
Simple, fast and efficient sample preparation approaches that allow high-throughput isolation of various compounds from complex matrices are highly desired in bioanalysis. Particularly sought are methods that can, without sacrificing time, easily remove matrix interferences capable of inducing ionization suppression/enhancement
Xiaoxia Mi et al.
Journal of chromatography. A, 1356, 54-63 (2014-07-16)
A method for analyzing 26 types of steroids in egg matrix was developed. The method used liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) in electrospray ionization mode (ESI). The procedure involved extraction with acetonitrile and removal of phospholipids by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service