Skip to Content
Merck
All Photos(1)

Key Documents

P3929

Supelco

Sigma Pseudo Corpse Scent

Formulation II

Synonym(s):

TM=["Sigma Pseudo"] Corpse Scent Formulation II

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

format

neat

Looking for similar products? Visit Product Comparison Guide

Application

For post-putrification detection
Sigma pseudo corpse scent may be used as an effective artificial scent in training cadaver dogs. The volatile compounds of its decomposition have been analyzed using gas chromatography coupled with time of flight mass spectrometry (GC–TOFMS).
Canine training aids for the detection of corpses.

Legal Information

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sonja Stadler et al.
Journal of chromatography. A, 1255, 202-206 (2012-05-05)
Cadaver dogs are trained on a variety of materials, including artificial or pseudo scents. The chemical components of commercially available pseudo scents are not known, so their accuracy as a decomposition odour mimic and their effectiveness as a canine training

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service