Skip to Content
Merck
All Photos(3)

Documents

916498

Sigma-Aldrich

Grubbs Catalyst® M800

Umicore

Synonym(s):

(SIMes)2Ru(Ind)Cl2, Bis[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(3-phenyl-1H-inden-1-ylidene)ruthenium(II), Grubbs Catalyst® M80 (C975-bis)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H62Cl2N4Ru
CAS Number:
Molecular Weight:
975.10
UNSPSC Code:
12352103
NACRES:
NA.22

Quality Level

form

powder

mp

>300 °C

Legal Information

Product of Umicore

This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

Additional information available at www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service