Skip to Content
Merck
All Photos(1)

Key Documents

73816

Millipore

BG11 Broth

suitable for microbiology, 100x concentrate, sterile filtered, NutriSelect® Basic

Synonym(s):

Cyanobacteria BG-11 Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

Quality Level

sterility

sterile (Filtered and Aseptic Handled)

form

liquid

shelf life

limited shelf life, expiry date on the label

composition

calcium chloride dihydrate, 36.7 mg/L
citric acid, 5.6 mg/L
dipotassium hydrogen phosphate, 31.4 mg/L
disodium magnesium ETDA, 1 mg/L
ferric ammonium citrate, 6 mg/L
magnesium sulfate, 36 mg/L
sodium carbonate, 20 mg/L
sodium nitrate, 1.5 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

4.0-4.6 (25 °C, not adjusted)

application(s)

agriculture
environmental
food and beverages
life science and biopharma

microbiology

storage temp.

2-8°C

suitability

cyanobacteria

Application

BG-11 is an universal medium for the cultivation and maintenance of blue green algae (cyanobacteria), such as Synechocystis 6803; Nostoc sp. PCC 9104 and Nostoc sp. PCC 9025. It is used for studies of the photosystems, as well as to identify the primary targets of the photoprocess.

Preparation Note

Dissolve the medium concentrate 1:100 with sterile water. It is recommended to adjust pH, under aseptic conditions, with 1M NaOH to 7.1 or as desired. Add the choosen sterile trace element solution.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Liq. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Light dilution via wavelength management for efficient high-density photobioreactors?
Matthew D
Biotechnology and Bioengineering (2017)
Matthew D Ooms et al.
Biotechnology and bioengineering, 114(6), 1160-1169 (2017-02-07)
The spectral distribution of light influences microalgae productivity; however, development of photobioreactors has proceeded largely without regard to spectral optimization. Here, we use monochromatic light to quantify the joint influence of path length, culture density, light intensity, and wavelength on
Jennifer Wadsworth et al.
Astrobiology, 19(8), 995-1007 (2019-06-14)
The European Space Agency's EXPOSE facility, located on the outside of the International Space Station, was used to investigate the survival of cell aggregates of a cyanobacterium, Gloeocapsa sp., in space and simulated martian conditions for 531 days in low
Eleni Koutra et al.
Bioresource technology, 294, 122232-122232 (2019-10-15)
Digested effluents are usually deprived of the appropriate levels of organic carbon or macro- and micro-nutrients to effectively sustain microalgal growth. In this regard, Parachlorella kessleri was cultivated in an agro-waste digestate supplemented with different glucose concentrations, magnesium and trace

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service