Skip to Content
Merck
All Photos(2)

Key Documents

68562

Supelco

Sudan III

analytical standard

Synonym(s):

1-[4-(Phenylazo)phenylazo]-2-naphthol, Cerasin Red, Fat Ponceau G, Fat Soluble Sudan, Scarlet B, Solvent Red 23, Sudan G, Sudan Red BK, Tony Red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N4O
CAS Number:
Molecular Weight:
352.39
Colour Index Number:
26100
Beilstein:
931185
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

199 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Oc1ccc2ccccc2c1\N=N\c3ccc(cc3)\N=N\c4ccccc4

InChI

1S/C22H16N4O/c27-21-15-10-16-6-4-5-9-20(16)22(21)26-25-19-13-11-18(12-14-19)24-23-17-7-2-1-3-8-17/h1-15,27H/b24-23+,26-25+

InChI key

FHNINJWBTRXEBC-QSZPNPOGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sudan III is a synthetic industrial azo-dye belonging to the group of sudan dyes, commonly used in plastics, oils, polishes and waxes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sudan III may be used as an analytical reference standard for the quantification of the analyte in food products using different chromatography techniques.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sampson Saj Andoh et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(7), 1049-1060 (2020-02-23)
Due to the proven carcinogenicity of Sudan III and IV dyes, they are considered global public health issues. They are banned in all forms as food colourants. We propose the monitoring of simple and easy-to-measure optical properties of palm oils
Analysis of sudan I, sudan II, sudan III, and sudan IV in food by HPLC with electrochemical detection: Comparison of glassy carbon electrode with carbon nanotube-ionic liquid gel modified electrode.
Chailapakul O
Food Chemistry, 109(4), 876-882 (2008)
Development and in-house validation of a liquid chromatography?electrospray?tandem mass spectrometry method for the simultaneous determination of Sudan I, Sudan II, Sudan III and Sudan IV in hot chilli products
Calbiani.F, et al.
Journal of Chromatography A, 1042(1-2), 123-130 (2004)
Desiree Van Haute et al.
International journal of pharmaceutics, 569, 118603-118603 (2019-08-12)
Conventional quantitation of drug content in the liposome formulation involves the breakdown of bulk liposomes, which ignores details on the distribution of the active pharmaceutical ingredient (API) and excipients in liposomes of different sizes. The objective of this study is
Eun Seong Lee et al.
Colloids and surfaces. B, Biointerfaces, 82(1), 190-195 (2010-09-21)
Pluronics with different structural compositions and properties are used for several applications, including drug delivery systems. We developed a binary mixing system with two Pluronics, L121/P123, as a nano-sized drug delivery carrier. The lamellar-forming Pluronic L121 (0.1 wt%) was incorporated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service