Skip to Content
Merck
All Photos(2)

Key Documents

32842

Supelco

Mebendazole-d3

VETRANAL®, analytical standard

Synonym(s):

5-Benzoyl-2-benzimidazolecarbamic acid methyl-d3 ester, Methyl-d3 N-(5-benzoyl-1H-benzimidazol-2-yl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D3H10N3O3
CAS Number:
Molecular Weight:
298.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])OC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccccc3

InChI

1S/C16H13N3O3/c1-22-16(21)19-15-17-12-8-7-11(9-13(12)18-15)14(20)10-5-3-2-4-6-10/h2-9H,1H3,(H2,17,18,19,21)/i1D3

InChI key

OPXLLQIJSORQAM-FIBGUPNXSA-N

General description

Mebendazole-d3 is a deuterated compound of anthelmintic drug, mebendazole.

Application

Mebendazole-d3 may be used as an internal standard for the determination of mebendazole in milk and animal muscle tissue samples by ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS). It can also be used as an internal standard for the determination of levamisole and aminorex in biological samples by liquid chromatography hybrid quadrupole time-of-flight high-resolution mass spectrometry (LC-QTOF-HRMS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue method for identification and quantification of avermectins, benzimidazoles and nitroimidazoles residues in bovine muscle tissue by ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) using a QuEChERS approach.
da Silva GR, et al.
Talanta, 171(27), 307-320 (2017)
Ji Sun Lee et al.
PloS one, 12(1), e0169597-e0169597 (2017-01-14)
A confirmatory and quantitative method of liquid chromatography-tandem mass spectrometry (LC-MS/MS) for the determination of mebendazole and its hydrolyzed and reduced metabolites in pork, chicken, and horse muscles was developed and validated in this study. Anthelmintic compounds were extracted with
Metabolism of levamisole and kinetics of levamisole and aminorex in urine by means of LC-QTOF-HRMS and LC-QqQ-MS.
Hess C, et al.
Analytical and Bioanalytical Chemistry, 405(12), 4077-4088 (2013)
Determination of anthelmintic drug residues in milk using ultra high performance liquid chromatography-tandem mass spectrometry with rapid polarity switching.
Whelan M, et al.
Journal of Chromatography A, 1217(27), 4612-4622 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service